Learn More
UVP Reciprocating Shaker Tray
For use in UVP hybridisation ovens
Brand: UVP 98-0067-02
Code : 04
Additional Details : Weight : 2.50000kg
Description
Provides accurate and consistent temperature control, variable rotation speed and come in various sizes. Some units incorporate other functions to increase free lab space. Controls are operated by keypad, rocker switch and dial.HB-500
- A desktop sized unit appropriate for start-up labs or labs with low level usage requirement.
HB-1000
- A standard sized unit designed for busy labs. The units can be stacked to maximise working space.
HL-2000
- HB-1000 oven and CX-2000 crosslinker combined in one space saving unit.
HM-4000
- Includes two compartments that operate independently of each other to provide a variety of motions.
The UVP hybridisation oven family are suitable for Southern, Northern and in situ hybridisation.
Specifications
Reciprocating Shaker Tray |