missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Coated Receiving Flask

Product Code. 10147701 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
3000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10147701 250 mL Each
10351442 100 mL Each
10438781 500 mL Each
10381532 1000 mL Each
10779562 3000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10147701 Supplier Heidolph™ Supplier No. 5148200002
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with rotary evaporators

  • With S 35/20
  • With transparent plastic coating
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Glass
Type Receiving Flask
For Use With (Equipment) Rotary evaporators
Material (Coating) Transparent plastic
Standard Taper Joint Number Center 35/20
Volume (Metric) 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.