Learn More
Invitrogen™ RECA-1 Monoclonal Antibody (HIS52), eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-0360-82
Description
Description: The HIS52 monoclonal antibody reacts with a molecule expressed on the surface of all rat endothelial cells, also known as Rat Endothelium Cell Antigen-1 (RECA-1). The exact nature of HIS52 antigen is, however, not known. HIS52 does not cross react with various species tested other than rat. This antibody is a very useful tool as a specific phenotypic marker for rat endothelium. Applications Reported: The HIS52 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. No reactivity of the antibody was seen when applied in immunoblotting of PAGE-run lysates from endothelial cell cultures and stromal cell preparations. Applications Tested: The HIS52 antibody has been tested by immunohistochemistry of rat frozen sections and can be used at 1-10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
Specifications
| RECA-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| HIS52 | |
| Unconjugated | |
| Reca1 | |
| Affinity chromatography | |
| RUO | |
| Rat | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.