missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RecA-1 Antibody (HIS52) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-64647JF669
This item is not returnable.
View return policy
Description
RecA-1 Monoclonal antibody specifically detects RecA-1 in Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Frozen),Immunofluorescence
Specifications
| RecA-1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| RecA protein, RecA1, RecA2, recaLP, Recombinase A | |
| Stromal cells from rat lymph node | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence | |
| HIS52 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction