Learn More
REC8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10793-1-AP-150UL
 View more versions of this product
Product Code. 16847413
  • £349.00 / 150µL
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human REC8. This antibody can recognize the monomeric, non-phosphorylated form (63-70 kDa) of REC8 and other forms(80-100 kDa) which represent either post transla
This gene encodes a member of the kleisin family of SMC (structural maintece of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene.
Specifications
Specifications
REC8
Polyclonal
Unconjugated
REC8
Cohesin Rec8p, HR21spB, REC8, REC8 homolog (yeast), REC8L1, Rec8p
Rabbit
Antigen Affinity Chromatography
RUO
56739, 9985
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95072, Q8C5S7
Rec8
REC8 Fusion Protein Ag1250
150 μL
Primary
Human, Mouse