missing translation for 'onlineSavingsMsg'
Learn More

REC8 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18692452 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18692452 0.02 mL 0.02mL
18663282 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18692452 Supplier Novus Biologicals Supplier No. NBP2951580.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

REC8 Polyclonal antibody specifically detects REC8 in Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen REC8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias Cohesin Rec8p, cohesion rec8p, HR21spB, meiotic recombination and sister chromatid cohesion phosphoprotein of therad21p family, meiotic recombination protein REC8 homolog, meiotic recombination protein REC8-like 1, MGC950, REC8 homolog (yeast), REC8L1human homolog of rad21, S. pombe, REC8-like 1, REC8-like 1 (yeast), Rec8p, recombination and sister chromatid cohesion protein homolog
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 194-561 of mouse REC8 (NP_064386.2). EVITLQEAEPIRMLQIEGEQDLPEISRGDLELLIAEKDDAILLEERQRGRLLRQRRASLPLDESREEPRALEGAGLVSALSPPAPAQVEGIQEALPGQVFPPEVQKMTGWEPGALLTEVTPPQELRLPAPPSTEKRLPSLQRPLPRRHRRRQLLFWDKETQISREKFEEQLQTGAHCWEYPVAQPPKRMLTSPAELFRTPTLSGWLPPELLGLWTHCAQVPQRMLRQRPQLETEETVEEERAADEEERRKTEALSEIEVLREAQEPSGPLMLSSELSLEAAEDEKSRTSLIPPEWWAWSEEGQPEPPALPMLPELPEVPMEMPPRPELSSEAVLRAVALKLQANKELDFSSLVPPLSPRKLASRVFYL
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9985
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.