missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
REC8 Monoclonal antibody specifically detects REC8 in Human samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Antigen | REC8 |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 2G3 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH04159 |
| Gene Alias | Cohesin Rec8p, cohesion rec8p, HR21spB, meiotic recombination and sister chromatid cohesion phosphoprotein of therad21p family, meiotic recombination protein REC8 homolog, meiotic recombination protein REC8-like 1, MGC950, REC8 homolog (yeast), REC8L1human homolog of rad21, S. pombe, REC8-like 1, REC8-like 1 (yeast), Rec8p, recombination and sister chromatid cohesion protein homolog |
| Host Species | Mouse |
| Immunogen | REC8L1 (AAH04159.1, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MFYYPNVLQRHTGCFATIWLAATRGSRLVKREYLRVNVVKTCEEILNYVLVRVQPPQPGLPRPRFSLYLSAQLQIGVIRVYSQQCQYLVEDIQHILERLHRAQLQIRIDM |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?