missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ REBM™ Renal Epithelial Cell Growth Basal Medium
Description
- Basal medium.
- Can be used with CC-4127(renal cell growth medium SingleQuots™ Supplements and Growth Factors).
- The full REGM™ BulletKit™ consisting of basal medium and supplements can be purchased together using the catalog number CC-3190.
- REGM™ Renal Epithelial Cell Growth Medium BulleKit™ is formulated to maintain and expand most renal epithelial cells.
- REGM™ contains 0.5% serum.
Specifications
Specifications
| Cell Type | Epithelial Cells |
| Product Type | Basal Medium |
| Quantity | 500 mL |
| pH | 7.4 to 7.6 |
| Color | Red |
| Content And Storage | Store in dark at 2 to 8°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?