missing translation for 'onlineSavingsMsg'
Learn More

Rebaudioside C, MedChemExpress

Product Code. 16459086
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459086 100 mg 100mg
16441418 5 mg 5mg
16408998 10 mg 10mg
16404847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16459086 Supplier Medchem Express Supplier No. HYN0467
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rebaudioside C(Dulcoside B) is used as natural sweeteners to diabetics and others on carbohydrate-controlled diets.

Rebaudioside C(Dulcoside B) is used as natural sweeteners to diabetics and others on carbohydrate-controlled diets.

Specifications

Chemical Name or Material Rebaudioside C
CAS 63550-99-2
Color White
Purity Grade Notes Research
Molecular Formula C44H70O22
Quantity 100 mg
Synonym Dulcoside B
Solubility Information DMSO : ≥ 40 mg/mL (42.06 mM)
SMILES O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC([C@@]2(C)[C@](CC[C@]3(CC4=C)[C@@]5([H])CC[C@]4(O[C@]6([H])O[C@H](CO)[C@@H](O)[C@H](O[C@]7([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H]6O[C@@]8([H])[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O8)C3)([H])[C@@]5(C)CCC2)=O)[C@@H]1O
Molecular Weight (g/mol) 951.01
Formula Weight 951.01
Percent Purity 98.21%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.