missing translation for 'onlineSavingsMsg'
Learn More

Rebaudioside A, MedChemExpress

Product Code. 16472376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472376 500 mg 500mg
16449086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472376 Supplier Medchem Express Supplier No. HYN0466
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Rebaudioside A is a steviol glycoside, α-glucosidase inhibitor with IC50 of 35.01 μg/ml.

Rebaudioside A is a steviol glycoside, α-glucosidase inhibitor with IC50 of 35.01 μg/ml.

Specifications

Chemical Name or Material Rebaudioside A
CAS 58543-16-1
Color White
Purity Grade Notes Research
Molecular Formula C44H70O23
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (103.41 mM)
SMILES C[C@]12[C@@]3([H])[C@@](C4)(CC[C@]1([H])[C@@](C)(CCC2)C(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)CO)=O)C[C@@](C4=C)(O[C@@]6([H])[C@@H]([C@H]([C@H](O)[C@@H](CO)O6)O[C@]7([H])O[C@@H]([C@@H](O)[C@H](O)[C@H]7O)CO)O[C@]8([H])O[C@@H]([C@@H](O)[C@H](O)[C@H]8O)CO)CC3
Molecular Weight (g/mol) 967.01
Formula Weight 967.01
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.