missing translation for 'onlineSavingsMsg'
Learn More

Rebastinib, MedChemExpress

Product Code. 16461667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461667 50 mg 50mg
16448207 5 mg 5mg
16405498 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16461667 Supplier Medchem Express Supplier No. HY13024
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.

Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.

Specifications

Chemical Name or Material Rebastinib
CAS 1020172-07-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H28FN7O3
Quantity 50 mg
Synonym DCC-2036
Solubility Information DMSO : 50 mg/mL (90.32 mM; ultrasonic and warming and heat to 80°C)
SMILES CC(C)(C)C1=NN(C(NC(NC2=C(F)C=C(OC3=CC(C(NC)=O)=NC=C3)C=C2)=O)=C1)C4=CC=C5C(C=CC=N5)=C4
Molecular Weight (g/mol) 553.59
Formula Weight 553.59
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.