missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ Reagents for Nitrogen-Ammonia Powder Pillow Set
Description
Premeasured powder reagents in individual packages for determination of Ammonia Nitrogen. Hach™ Reagents for Nitrogen-Ammonia Powder Pillow Set include Ammonia Cyanurate and Ammonia Salicylate reagents that can be purchased together or used separately.
- Ready-to-use, premeasured powder reagents in individual sealed packets
- For Ammonia Nitrogen determination by the Salicylate Method (Hach Method 55)
- Ammonia Cyanurate and Ammonia Salicylate powder pillows for 100 tests
- Both sets are contained in the Hach Nitrogen-Ammonia Powder Pillow Reagent Set (man. no. 2668000)
Specifications
Specifications
| Analytes | Ammonia Nitrogen |
| Color | White |
| Format | Individual Packet |
| For Use With (Application) | Ammonia Nitrogen determination |
| For Use With (Equipment) | All except DR/4000, DR/3000, DR/2000, DR 900 |
| Includes | Ammonia Cyanurate Reagent |
| No. of Tests | 100 |
| Quantity | 100 Pack |
| Range | 0.01 to 0.50mg/L NH3-N |
| Sample Size | 10 mL |
| Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?