missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ Reagents for Nitrogen-Ammonia Powder Pillow Set
Click to view available options
Includes:
Ammonia Cyanurate Reagent
Ammonia Salicylate Reagent
Unit Size:
Each
Pack of 100
Description
Premeasured powder reagents in individual packages for determination of Ammonia Nitrogen. Hach™ Reagents for Nitrogen-Ammonia Powder Pillow Set include Ammonia Cyanurate and Ammonia Salicylate reagents that can be purchased together or used separately.
- Ready-to-use, premeasured powder reagents in individual sealed packets
- For Ammonia Nitrogen determination by the Salicylate Method (Hach Method 55)
- Ammonia Cyanurate and Ammonia Salicylate powder pillows for 100 tests
- Both sets are contained in the Hach Nitrogen-Ammonia Powder Pillow Reagent Set (man. no. 2668000)
Specifications
Specifications
| Analytes | Ammonia Nitrogen |
| Color | White |
| Format | Individual Packet |
| For Use With (Application) | Ammonia Nitrogen determination |
| For Use With (Equipment) | All except DR/4000, DR/3000, DR/2000, DR 900 |
| Includes | Ammonia Salicylate Reagent |
| No. of Tests | 100 |
| Quantity | 100 Pack |
| Range | 0.01 to 0.50mg/L NH3-N |
| Sample Size | 10 mL |
| Show More |
Safety and Handling
Product Identifier
- Ammonia Salicylate Reagent
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : sodium salicylate, sodium nitroferricyanide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction