missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reagent Reservoirs
GREENER_CHOICE

Product Code. 10481121
Change view
Click to view available options
Capacity (Metric):
25 mL
60 mL
100 mL
Material:
Polypropylene
Polystyrene
Sterility:
Non-sterile
Packaging:
10 Bags of 10
Bulk
Unit Size:
Pack of 100
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material Sterility Packaging Unit Size
10481121 60 mL Polypropylene Non-sterile Bulk Pack of 5
11475748 25 mL Polystyrene Non-sterile 10 Bags of 10 Pack of 100
10125707 100 mL Polystyrene Non-sterile 10 Bags of 10 Pack of 100
11405758 25 mL Polystyrene Non-sterile Bulk Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10481121 Supplier Thermo Scientific™ Supplier No. 9510027
Curiosity Goes Beyond
Use promo code 27966
View all promotions
In Stock
Add to basket
Edge
Add to basket

Choose these reagent reservoirs, designed to provide a variety of solutions to suit all of your pipetting procedures.

Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.

Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.

Benefits

  • Trough within a trough maximizes the amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • 25mL Divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 2 oz.
Color White
Capacity (Metric) 60 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
Packaging Bulk
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.