missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Reagent Reservoir Dual - 25 mL
BIOPHARMA

Product Code. 16374862 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Packaging:
1/bag, 100 bags
5/bag, 40 bags
Unit Size:
Pack of 100
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
16374862 1/bag, 100 bags Pack of 100
16364862 5/bag, 40 bags Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16374862 Supplier Heathrow Scientific™ Supplier No. 120652
Quantity Price
1 £176.00 / Pack of 100
6 £158.40 / Pack of 100
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures

  • Divided reservoir design allows pipetting of two different regents or maximizes recovery of using only single channel and small amount of reagent
  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sterile, sealed in bags and electron beam sterilized
  • DNase, RNase and Pyrogen free
  • Sealed in easy-to-open bags. No scissors needed to open
  • Quality certificate available on request
  • Disposable
  • Design Patent Pending

Specifications

Capacity (English) .85 oz
Certifications/Compliance Dnase, Rnase & Pyrogen Free
Coating None
Color White
Description Single wrap
Capacity (Metric) 25 mL
Dimensions (L x W x H) 4.1 x 1 x 0.8 in
Material PVC
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) Reagent
Height (English) 0.8 in
Height (Metric) 2.1 cm
Includes Reservoir
Length (English) 4 in
Length (Metric) 10.2 cm
Max. Temperature (English) 104°F Operating
Max. Temperature (Metric) 40°C Operating
No. per Case 600
No. per Pack 100
Packaging 1/bag, 100 bags
Quantity 100
Volume (Metric) Well 25 mL
Width (English) 1 in
Width (Metric) 2.5 cm
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Conical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.