missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Reagent Diluent Concentrate 3 (5X)

Product Code. 30021769
Change view
Click to view available options
Quantity:
10 x 21 mL
Unit Size:
210mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30021769 10 x 21 mL 210mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021769 Supplier R&D Systems™ Supplier No. DY004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5X Concentrated 25% Tween 20 solution in PBS for DuoSets

10 vials (21mL/vial) of a 25% Tween 20 solution in PBS. The reagent diluent will require additional components for some DuoSets. Refer to the product-specific package. insert for more information.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Quantity 10 x 21 mL
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.