missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Reagent Diluent Concentrate 2

Product Code. 18754394
Change view
Click to view available options
Quantity:
5 x 21 mL
Unit Size:
105mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18754394 5 x 21 mL 105mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18754394 Supplier R&D Systems™ Supplier No. DY995

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

10X Concentrated 10% BSA Solution for DuoSets

  • 10% BSA solution in a concentrated formula which can be diluted to make a working solution at a range of final BSA concentrations suitable for the assay being run
  • Make all solutions fresh or filter sterilize and store for up to 1 week at 2° to 8°C
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Quantity 5 x 21 mL
compliance-icons
Product Identifier
  • Reagent Diluent Concentrate 2 (10X)
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Mixture of 5-Chloro-2-Methyl-4- Isothiazolin-3-One and 2-Methyl-4-Isothiazolin-3- One
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.