missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ In Situ Hybridization (ISH) Blocking Solution (5X)

Product Code. 17123773
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17123773 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17123773 Supplier Invitrogen™ Supplier No. R37620
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Minimizes non-specific background staining in chromogenic in situ hybridization (CISH) or fluorescence in situ hybridization (FISH) procedures

  • ReadyProbes In Situ Hybridization (ISH) Blocking Solution minimizes non-specific background staining in chromogenic in situ hybridization (CISH) or fluorescence in situ hybridization (FISH) procedures
  • It is provided as a 5X solution that is diluted to 1X with distilled or deionized water
  • After the labeled DNA or RNA probe hybridization and wash steps, the sample is incubated with 1X Ready Probes ISH Blocking Solution for 30–60 minutes
  • 1X ReadyProbes ISH Blocking Solution can also be used as diluent for antibodies, avidin, streptavidin, or biotin during incubation steps.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Provided as 100-mL solution at 5X concentration in amber HDPE bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Concentration 5X
Product Line ReadyProbes
Product Type In Situ Hybridization Blocking Solution
Quantity 100 mL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.