missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Hydrophobic Barrier Pap Pen

Product Code. 16660484
Change view
Click to view available options
Quantity:
2 Pens
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16660484 2 Pens Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16660484 Supplier Invitrogen™ Supplier No. R37622

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip.

The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip. By lightly drawing a rectangle or circle around the tissue section or cells on the microscope slide, aqueous incubation reagents can be kept inside the circle or rectangle. Save precious reagents by incubating with less volume. The ReadyProbes Hydrophobic Barrier Pap Pen can also be used to keep reagents separate when more than one tissue section is mounted on a microscope slide.

The light blue barrier line is heat resistant and compatible with most immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) reagents, including detergent-containing reagents. It is insoluble in alcohol and acetone, but can be removed with clearing agent (e.g., xylene).

To use, dab the nib of the pen on a paper towel to make sure the flow of the hydrophobic fluid is uninterrupted. Lightly trace a circle or rectangle around the tissue/cells 2–4 times with a thick line, while keeping few millimeters distance from the edge of the tissue/cells. Let dry for 20–30 minutes at room temperature. Repeat the process, if needed.

TRUSTED_SUSTAINABILITY

Specifications

Type Hydrophobic Barrier Pap Pen
Product Line ReadyProbes
Content And Storage A set of 2 pens.

Store (at room temperature.

When stored as directed, product can be used for 6 months after receipt.

)
Includes 2 Pens
Quantity 2 Pens
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.