missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ready-to-Go™ DNA Labeling Beads (-dCTP)

Product Code. 10607475 Shop All Cytiva Products
Click to view available options
:
Sufficient for 20 rxns
This item is not returnable. View return policy

Product Code. 10607475

Brand: Cytiva 27924001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Labeling beads (from a buffered aqueous solution containing dATP, dGTP, dTTP, random nonamers [d(N)9] and Klenow fragment) and control DNA bead

Ambient-temperature-stable, single-dose reaction spheres for labeling DNA to specific activities >109dpm/μg in as little as 5 min. using labeled dCTP

  • Beads are predispensed in individual reaction tubes to eliminate the potential for cross-contamination
  • Beads reconstitute immediately upon addition of water or buffer to a final reaction volume of 50μL
  • DNA can be labeled in solution or in the presence of low melting point agarose
  • Reaction mixtures efficiently incorporate 32P- or 35S-labeled dCTP
  • Each reaction is capable of labeling between 10ng and 1μg of linearized DNA

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) DNA labeling bead can be used with Either 32P or 35S labeled dCTP
Includes Labeling beads and control DNA bead
No. of Reactions 20
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.