missing translation for 'onlineSavingsMsg'
Learn More

Ready-Made LB Agar Powder

Product Code. 15885368 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 15885368

Brand: Thermo Scientific J75851.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For propagation of bacteria

pH(3.5%, H2O): 7.0 ± 0.2

Contents:

  • Casein Peptone: 10gm/L
  • Yeast Extract: 5gm/L
  • Sodium Chloride: 5gm/L
  • Agar: 15gm/L
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Agar
For Use With (Application) Propagation of Bacteria
Packaging Type Plastic Bottle
Quantity 1 kg
What instructions should I use to prepare Ready-Made LB Agar Powder (Cat. No. J75851.A1)?

We recommend dissolving 40 grams of the medium in 1 L of purified water until evenly dispersed. Heat with repeated stirring and boil for one minute to dissolve completely. Distribute into bottles for autoclaving and autoclave at 121 degrees C for 15 minutes.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.