missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
At the visible spectrum, Cibacron Brilliant Red 3B-A exhibits a ?max and ?min of 567nm and 501nm respectively. Studies suggest that the minimum structure required for this dye to bind hexokinase is 1-amino-8-naphthol-3,6-disulfonic acid.
Specifications
Specifications
| CAS | 17681-50-4 |
| Molecular Formula | C32H19ClN8Na4O14S4 |
| Quantity | 5 g |
| Synonym | reactive red 4, cibacron brilliant red 3b-a, tetrasodium 5-benzamido-3-e-2-5-4-chloro-6-4-sulfonatophenyl amino-1,3,5-triazin-2-yl amino-2-sulfonatophenyl diazen-1-yl-4-hydroxynaphthalene-2,7-disulfonate |
| Solubility Information | Soluble in water. Very slightly soluble in ethanol and methanol |
| InChI Key | HMBFFLCYQUKBIW-UHFFFAOYSA-J |
| SMILES | C1=CC=C(C=C1)C(=O)NC2=C3C(=CC(=C2)S(=O)(=O)[O-])C=C(C(=NNC4=C(C=CC(=C4)NC5=NC(=NC(=N5)NC6=CC=C(C=C6)S(=O)(=O)[O-])Cl)S(=O)(=O)[O-])C3=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+] |
| IUPAC Name | tetrasodium;5-benzamido-3-[[5-[[4-chloro-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate |
| Molecular Weight (g/mol) | 995.195 |
| PubChem CID | 44134921 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?