missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Reaction Vessels for Abbot Architect Series Analyzers

Product Code. 31922854
Change view
Click to view available options
Quantity:
2000
Unit Size:
Pack of 10000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31922854 2000 Pack of 10000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31922854 Supplier Globe Scientific Supplier No. 5105
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed specifically for use with Abbot Architect Series Analyzers. Globe Scientific Reaction Vessel for Abbot Architect Series Analyzers are produced from polypropylene (PP).

  • For use with Abbot Architect Series Analyzers
  • Designed specifically to work with these products
  • Polypropylene construction, natural color

Specifications

Type Reaction Vessel
Color Natural
Quantity 2000
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.