Learn More
Elma™ Reaction Tube Holders
Product Code. 17873094
Change view
Click to view available options
Model:
100/120
150/300
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17873094 100/120 Each
17871357 150/300 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17873094 Supplier Elma™ Supplier No. 1112158
Ensure that reaction tubes can be securely positioned and arranged in the Elma Basket.
  • Optimum positioning of the items inside the bath is also a prerequisite for achieving the best possible cleaning results
  • Allows items to be individually positioned
  • Lets you clearly structure your cleaning process
  • Prevents scratches on the tank floor
  • Holders are available for various applications
  • Special solutions are available (e.g. for conical flasks)
Specifications
Type Reaction Tube Holder
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Model 100/120