missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ rea-sol™ Rapid Cleaning Agent

Product Code. 10748022
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10748022 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10748022 Supplier Hirschmann™ Supplier No. 9720105
  • ONLINE EXCLUSIVE PRICE
    £169.00
    £147.59 / 5 L
    Save £21.41
    12% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For rapid, automatic and careful cleaning of materials such as glass, porcelain, rubber, plastics and metals. Hirschmann Instruments™ rea-sol™ Rapid Cleaning Agent is ideal for the laboratory in removing oil, grease, wax, resin, silicone, blood, protein and serum residues.

  • With anti-corrosion additives
  • Sensitive tests not influenced
  • Free of chlorine and caustic alkalis
  • Environmentally friendly and non-toxic

Specifications

Product Type Cleaning Agent
Product Line Rea Sol
Concentration 1%
Container Type Canister
For Use With (Application) Anti Corrosion Additives
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • rea-sol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.