missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20156-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein involved in the cellular response to cisplatin, a drug commonly used in chemotherapy. The protein encoded by this gene contains two motifs: a motif found in RAD52, a protein that functions in DNA double-strand breaks and homologous recombination, and an RNA recognition motif (RRM) that is not found in RAD52. The RAD52 motif region in RAD52 is important for protein function and may be involved in DNA binding or oligomerization. Alternatively spliced transcript variants encoding different isoforms have been reported.Specifications
| RDM1 | |
| Polyclonal | |
| Unconjugated | |
| RDM1 | |
| RAD52 homolog B, RAD52 motif 1, RAD52B, RDM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 201299 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NG50 | |
| RDM1 | |
| RDM1 Fusion Protein Ag14077 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction