missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDM1 Mouse anti-Human, Unconjugated, Clone: 1C10B4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RDM1 |
|---|---|
| Clone | 1C10B4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
16829255
|
Proteintech
66941-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16819255
|
Proteintech
66941-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RDM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RDM1. This antibody recognizes human, mouse, rat antigen. The RDM1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
| RDM1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 201299 | |
| Primary |
| 1C10B4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG2a |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit