missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDM1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95218-0.1ml
This item is not returnable.
View return policy
Description
RDM1 Polyclonal antibody specifically detects RDM1 in Mouse samples. It is validated for Western Blot
Specifications
| RDM1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| MGC33977, RAD52 homolog B, RAD52 homolog B (S. cerevisiae), RAD52 motif 1, RAD52 motif-containing protein 1, RAD52B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 125-284 of human RDM1 (NP_663629.1). NGCSKRIIKLQELSDLEERENEDSMVPLPKQSLKFFCALEVVLPSCDCRSPGIGLVEEPMDKVEEGPLSFLMKRKTAQKLAIQKALSDAFQKLLIVVLESGKIAVEYRPSEDIVGVRCEEELHGLIQVPCSPWKQYGQEEEGYLSDFSLEEEEFRLPELD | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 201299 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction