missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16283-1-AP-20UL
This item is not returnable.
View return policy
Description
Exhibits an oxidoreductive catalytic activity towards retinoids. Most efficient as an NADPH-dependent retinal reductase. Displays high activity toward 9-cis and all-trans-retinol. No steroid dehydrogenase activity detected.Specifications
| RDH14 | |
| Polyclonal | |
| Unconjugated | |
| RDH14 | |
| Alcohol dehydrogenase PAN2, PAN2, RDH14, Retinol dehydrogenase 14, SDR7C4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105014, 500629, 57665 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ERI6, Q9HBH5 | |
| Rdh14 | |
| RDH14 Fusion Protein Ag9386 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction