missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH14 Antibody (OTI1A7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73847UV
This item is not returnable.
View return policy
Description
RDH14 Monoclonal specifically detects RDH14 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| RDH14 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RDH14 | |
| Full length recombinant protein of human RDH14 (NP_065956) produced in HEK293T cells. | |
| 0.1 mL | |
| 57665.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI1A7 | |
| Western Blot, Flow Cytometry | |
| Alcohol dehydrogenase PAN2, EC 1.1.1.-, PAN2retinol dehydrogenase 14, pancreas protein 2, retinol dehydrogenase 14 (all-trans and 9-cis), retinol dehydrogenase 14 (all-trans/9-cis/11-cis), SDR7C4, short chain dehydrogenase/reductase family 7C, member 4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction