missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11077-1-AP-20UL
This item is not returnable.
View return policy
Description
RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.Specifications
| RDH11 | |
| Polyclonal | |
| Unconjugated | |
| RDH11 | |
| ARSDR1, CGI 82, HCBP12, MDT1, PSDR1, RALR1, RDH11, Retinal reductase 1, Retinol dehydrogenase 11, SCALD, SDR7C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17252, 362757, 51109 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8TC12, Q9QYF1 | |
| Rdh11 | |
| RDH11 Fusion Protein Ag1485 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction