missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35847-20ul
This item is not returnable.
View return policy
Description
RDH11 Polyclonal antibody specifically detects RDH11 in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RDH11 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Androgen-regulated short-chain dehydrogenase/reductase 1, ARSDR1CGI82, EC 1.1.1.300, HCBP12, HCV core-binding protein HCBP12, MDT1, prostate short-chain dehydrogenase reductase 1, Prostate short-chain dehydrogenase/reductase 1, PSDR1FLJ32633, RALR1, Retinal reductase 1, retinol dehydrogenase 11, retinol dehydrogenase 11 (all-trans and 9-cis), retinol dehydrogenase 11 (all-trans/9-cis/11-cis), SCALD, SDR7C1, short chain dehydrogenase/reductase family 7C, member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-145 of human RDH11 (NP_057110.3).,, Sequence:, LACRDVEKGELVAKEIQTTTGNQQVLVRKLDLSDTKSIRAFAKGFLAEEKHLHVLINNAGVMMCPYSKTADGFEMH | |
| 20 μL | |
| Vision | |
| 51109 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction