missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RD3 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | RD3 |
|---|---|
| Concentration | 850 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30233187
|
Proteintech
14855-1-AP-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233325
|
Proteintech
14855-1-AP-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The RD3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RD3. This antibody recognizes human, mouse, rat antigen. The RD3 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Spezifikation
| RD3 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| C1orf36; LCA12 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 850 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 343035 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts