missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCOR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55173-1-AP-150UL
This item is not returnable.
View return policy
Description
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.Specifications
| RCOR3 | |
| Polyclonal | |
| Unconjugated | |
| RCOR3 | |
| KIAA1343, RCOR3, REST corepressor 3, RP11 318L16.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55758 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9P2K3 | |
| RCOR3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction