missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCOR2 Polyclonal Antibody

RCOR2 Polyclonal Antibody
Brand: Thermo Scientific PA554477
This item is not returnable.
View return policy
Description
RCOR2 Polyclonal Antibody for ICC/IF, IHC
Specifications
| RCOR2 | |
| Polyclonal | |
| Unconjugated | |
| RCOR2 | |
| 1A13; AW122124; co-repressor of Rest; CoREST; M-CoREST; Rcor; Rcor1; Rcor2; RE1-silencing transcription factor (REST) co-repressor 1; RE1-silencing transcription factor (REST) co-repressor 2; REST corepressor 2; REST co-repressor 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283248 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZ40 | |
| RCOR2 | |
| Recombinant protein corresponding to Human RCOR2. Recombinant protein control fragment (Product #RP-93618). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction