missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCOR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107238
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 86%.
RCOR2 may act as a component of a corepressor complex that represses transcription.
Specifications
| RCOR2 | |
| Polyclonal | |
| Unconjugated | |
| RCOR2 | |
| 1A13; AW122124; co-repressor of Rest; CoREST; M-CoREST; Rcor; Rcor1; Rcor2; RE1-silencing transcription factor (REST) co-repressor 1; RE1-silencing transcription factor (REST) co-repressor 2; REST corepressor 2; REST co-repressor 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 104383 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q8C796 | |
| RCOR2 | |
| Synthetic peptide directed towards the C-terminal region of Mouse Rcor2. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction