missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCOR1 Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human RCOR1.
Brand: Abnova PAB29456.100uL
This item is not returnable.
View return policy
Description
The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression.[supplied by OMIM]
Sequence: SSDEEHGGGGMRVGPQYQAVVPDFDPAKLARRSQERDNLGMLVSpecifications
| RCOR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant human RCOR1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| RCOR1 | |
| Recombinant protein corresponding to human RCOR1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| COREST/KIAA0071/RCOR | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23186 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction