missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCOR1/CoREST Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-240F
This item is not returnable.
View return policy
Description
RCOR1/CoREST Polyclonal antibody specifically detects RCOR1/CoREST in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RCOR1/CoREST | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| KIAA0071CORESTRCORREST corepressor, Protein CoREST, REST corepressor 1 | |
| A synthetic peptide that maps to a region between residues 425 and the C-terminus (residue 482) of human REST (RE1-silencing transcription factor) corepressor using the numbering given in entry NP_055971.1 (GeneID 23186) | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 23186 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction