missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £434.00
Specifications
| Antigen | RCN2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231715
|
Novus Biologicals
NBP3-38457-100ul |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232115
|
Novus Biologicals
NBP3-38457-20ul |
20 μL |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RCN2 Polyclonal antibody specifically detects RCN2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| RCN2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 5955 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Calcium-binding protein ERC-55, E6BPreticulocalbin 2, EF-hand calcium binding domain (endoplasmic reticulumcalcium-binding protein, 55kD), ERC-55, ERC55E6-binding protein, reticulocalbin 2, EF-hand calcium binding domain, reticulocalbin-2, TCBP49 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-180 of human RCN2 (NP_002893.1).,, Sequence:, GKAEELHYPLGERRSDYDREALLGVQEDVDEYVKLGHEEQQKRLQAIIKKIDLDSDGFLTESELSSWIQMSFKHYAMQEAKQQFVEYDKNSDDTVTWDEYNIQMYDRVIDFDENTALDDAEEESFRKLHLKDKKRFEKANQDSGPGLSLEEFIAFEHP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title