missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RCN1 Protein

Product Code. 18206012 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
0.25 mg
Unit Size:
0.05mg
0.25mg
This item is not returnable. View return policy

Product Code. 18206012

Brand: Novus Biologicals™ NBP1453210.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 30-331 of Human RCN1 The Recombinant Human RCN1 Protein is derived from E. coli. The Recombinant Human RCN1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1mM DTT, 100mM NaCl
Gene ID (Entrez) 5954
Molecular Weight (g/mol) 40.4kDa
Purification Method Protein
Quantity 0.05 mg
Source Human
Immunogen MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSELEK PTVRKERVVR PDSELGERPP EDNQSFQYDH EAFLGKEDSK TFDQLTPDES KERLGKIVDR IDNDGDGFVT TEELKTWIKR VQKRYIFDNV AKVWKDYDRD KDDKISWEEY KQATYGYYLG NPAEFHDSSD HHTFKKMLPR DERRFKAADL NGDLTATREE FTAFLHPEEF EHMKEIVVLE TLEDIDKNGD GFVDQDEYIA DMFSHEENGP EPDWVLSERE QFNEFRDLNK DGKLDKDEIR HWILPQDYDH AQAEARHLVY ESDKNKDEKL TKEEILENWN MFVGSQATNY GEDLTKNHDE L
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name RCN1
Conjugate Unconjugated
Purity or Quality Grade >80%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt