missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558409
This item is not returnable.
View return policy
Description
Immunogen sequence: LGKIVDRIDN DGDGFVTTEE LKTWIKRVQK RYIFDNVAKV WKDYDRDKDD KISWEEYKQA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Reticulocalbin 1 is a calcium-binding protein located in the lumen of the ER. The protein contains six conserved regions with similarity to a high affinity Ca(+2)-binding motif, the EF-hand. High conservation of amino acid residues outside of these motifs, in comparison to mouse reticulocalbin, is consistent with a possible biochemical function besides that of calcium binding. In human endothelial and prostate cancer cell lines this protein localizes to the plasma membrane.
Specifications
| RCN1 | |
| Polyclonal | |
| Unconjugated | |
| RCN1 | |
| epididymis secretory protein Li 84; FLJ37041; HEL-S-84; PIG20; proliferation-inducing gene 20; Rca1; RCAL; Rcn; RCN1; reticulocalbin 1; reticulocalbin 1, EF-hand calcium binding domain; reticulocalbin-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19672, 5954 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q05186, Q15293 | |
| RCN1 | |
| Recombinant protein corresponding to Human RCN1. Recombinant protein control fragment (Product #RP-97267). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction