missing translation for 'onlineSavingsMsg'
Learn More

RCM-1, MedChemExpress

Product Code. 16447776
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447776 100 mg 100mg
16470458 5 mg 5mg
16437998 10 mg 10mg
16423726 25 mg 25mg
16403927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16447776 Supplier Medchem Express Supplier No. HY19979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RCM-1 is a forkhead box M1 (FOXM1) inhibitor with an EC50 of 0.72 μM in U2OS cells. RCM-1 blocks the nuclear localization and increased the proteasomal degradation of FOXM1. RCM-1 can be used for asthma and other chronic airway diseases research.

RCM-1 is a forkhead box M1 (FOXM1) inhibitor with an EC50 of 0.72 μM in U2OS cells. RCM-1 blocks the nuclear localization and increased the proteasomal degradation of FOXM1. RCM-1 can be used for asthma and other chronic airway diseases research.

Specifications

Chemical Name or Material RCM-1
CAS 339163-65-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H12N2OS4
Quantity 100 mg
Solubility Information DMSO : 16.67 mg/mL (39.26 mM; Need ultrasonic)
SMILES N#CC1=C(C2=CC=CS2)C=C(C3=CC=CS3)N=C1SCC(C4=CC=CS4)=O
Molecular Weight (g/mol) 424.58
Formula Weight 424.58
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.