missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576587
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
RCL1, which does not have cyclase activity, plays a role in 40S- ribosomal-subunit biogenesis in the early pre-rRNA processing steps at sites A0, A1 and A2 that are required for proper maturation of the 18S RNA.
Specifications
| RCL1 | |
| Polyclonal | |
| Unconjugated | |
| RCL1 | |
| 2310040A02Rik; Ab1-353; Ac1292; AI789745; C76567; HSPC338; Rcl1; RNA 3'-terminal phosphate cyclase-like protein; RNA cyclase homolog; RNA terminal phosphate cyclase like 1; RNA terminal phosphate cyclase-like 1; Rnac; RPC2; RPCL1; RTC2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10171, 309301, 59028 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9JJT0, Q9Y2P8 | |
| RCL1 | |
| Recombinant fusion protein of human RCL1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction