missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15096
This item is not returnable.
View return policy
Description
RCL1 Polyclonal specifically detects RCL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RCL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| RNA terminal phosphate cyclase-like 1,2310040A02Rik | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9Y2P8 | |
| RCL1 | |
| Recombinant protein encompassing a sequence within the center region of human RCL1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10171 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction