missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCL Recombinant Rabbit Monoclonal Antibody (23GB5180), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552811
This item is not returnable.
View return policy
Description
RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.
Specifications
| RCL | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DNPH1 | |
| 2'-deoxynucleoside 5'-phosphate N-hydrolase 1; 2'-deoxynucleoside 5'-phosphate N-hydrolase 1 {ECO:0000255; C6orf108; C76683; c-Myc-responsive protein RCL; c-Myc-responsive protein Rcl {ECO:0000255; Deoxyribonucleoside 5'-monophosphate N-glycosidase; dJ330M21.3; DNPH1; HAMAP-Rule:MF_03036}; Nucleoside 2-deoxyribosyltransferase domain containing protein RGD620382; putative c-Myc-responsive; Rcl; RGD620382; RP3-330M21.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10591 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5180 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43598 | |
| DNPH1 | |
| A synthesized peptide derived from human DNPH1 (145-174AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction