missing translation for 'onlineSavingsMsg'
Learn More

RCGD423, MedChemExpress

Product Code. 16483256
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483256 25 mg 25mg
16467597 5 mg 5mg
16464848 10 mg 10mg
16451107 50 mg 50mg
16465636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483256 Supplier Medchem Express Supplier No. HY114775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RCGD423 is a gp130 modulator, which prevents articular cartilage degeneration and promotes repair.

RCGD423 is a gp130 modulator, which prevents articular cartilage degeneration and promotes repair.

Specifications

Chemical Name or Material RCGD423
CAS 108237-91-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H11BrN2S
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (377.38 mM; Need ultrasonic)
SMILES BrC1=CC=C(C=C1)NC2=NC(C3=CC=CC=C3)=CS2
Molecular Weight (g/mol) 331.23
Formula Weight 331.23
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.