missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ RCGD 423

Description
RCGD 423 is a gp130 modulator. Binds gp130, promotes Myc expression, increases lactate production and activates downstream JAK/STAT signaling pathways in hair follicle stem cells. Also increases articular chondrocyte activation and facilitates joint repair by regulating cartilage growth and differentiation.
Specifications
Specifications
| Chemical Name or Material | N-(4-Bromophenyl)-4-phenyl-2-thiazolamine |
| CAS | 108237-91-8 |
| Quantity | 50 mg |
| Molecular Formula | C15H11BrN2S |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?