missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCCD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559294
This item is not returnable.
View return policy
Description
Immunogen sequence: PFIAVQPFPA LLDLPMGSDA VKASCGSRHT AVVTRTGELY TWGWGKYGQL GHEDTTSLDR PRRVEYFVDK QLQVKAVTCG PWNTYVYAVE KG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Plays a role in transcriptional repression of satellite repeats, possibly by regulating H3K36 methylation levels in centromeric regions together with KDM8 (PubMed:24981860). Possibly together with KDM8, is involved in proper mitotic spindle organization and chromosome segregation (PubMed:24981860). Plays a role in regulating alpha-tubulin deacetylation and cytoskeletal microtubule stability, thereby promoting cell migration and TGF-beta-induced epithelial to mesenchymal transition (EMT), potentially through the inhibition of KDM8 (PubMed:28455245).
Specifications
| RCCD1 | |
| Polyclonal | |
| Unconjugated | |
| RCCD1 | |
| RCC1 domain containing 1; RCC1 domain-containing protein 1; RCCD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NED2 | |
| RCCD1 | |
| Recombinant protein corresponding to Human RCCD1. Recombinant protein control fragment (Product #RP-98162). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido