Learn More
Invitrogen™ RCC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143953
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell. ICC/IF: MCF-7 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Required for completion of mitosis and cytokinesis. May function as a guanine nucleotide exchange factor for the small GTPase RAC1.
Specifications
| RCC2 | |
| Polyclonal | |
| Unconjugated | |
| RCC2 | |
| 2610510H01Rik; 2610529N02Rik; AA536646; AA675016; KIAA1470; mKIAA1470; protein RCC2; Protein RCC2 homolog; RCC1-like; RCC1-like protein TD-60; RCC2; regulator of chromosome condensation 2; RGD1309986; Td60; td-60; Telophase disk protein of 60 kDa; wu:fb36a03; zgc:77115 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55920 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q9P258 | |
| RCC2 | |
| E.coli-derived human RCC2 recombinant protein (Position: K293-L522). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.