missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599702
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RCC2.
Required for completion of mitosis and cytokinesis. May function as a guanine nucleotide exchange factor for the small GTPase RAC1.
Specifications
| RCC2 | |
| Polyclonal | |
| Unconjugated | |
| RCC2 | |
| 2610510H01Rik; 2610529N02Rik; AA536646; AA675016; KIAA1470; mKIAA1470; protein RCC2; Protein RCC2 homolog; RCC1-like; RCC1-like protein TD-60; RCC2; regulator of chromosome condensation 2; RGD1309986; Td60; td-60; Telophase disk protein of 60 kDa; wu:fb36a03; zgc:77115 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108911, 298594, 55920 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BK67, Q9P258 | |
| RCC2 | |
| A synthesized peptide derived from human RCC2(Accession Q9P258), corresponding to amino acid residues C437-E487. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction